ContactUsLinkComponent
MULTIPROBE REDOX ASSAY KIT CELL PERMEANT REAGENT DIHYDROETHIDIUM IS A FLUOROGENIC DYE THAT IS USEFUL FOR THE DETECTION OF REACTIVE OXYGEN SPECIES 1 * 1 KIT
Catalog # GENOBAQ048
Supplier:  G-Biosciences
MULTIPROBE REDOX ASSAY KIT CELL PERMEANT REAGENT DIHYDROETHIDIUM IS A FLUOROGENIC DYE THAT IS USEFUL FOR THE DETECTION OF REACTIVE OXYGEN SPECIES 1 * 1 KIT
Catalog # GENOBAQ048
Supplier:  G-Biosciences
Frequently Bought Together