ContactUsLinkComponent
Stericlin® White Line pouches, non-woven polypro / film, very resistant, suitable for STEAM 134°C, EO, FORM and PLASMA, cleanliness level 200 per IEST-STD-CC1246, 56x66cm, DIN EN ISO 11607, case of 5 packs x 50pcs 1 * 250 items
Catalog # VEPA3FKFB245185
Stericlin® White Line pouches, non-woven polypro / film, very resistant, suitable for STEAM 134°C, EO, FORM and PLASMA, cleanliness level 200 per IEST-STD-CC1246, 56x66cm, DIN EN ISO 11607, case of 5 packs x 50pcs 1 * 250 items
Catalog # VEPA3FKFB245185