Stericlin® White Line pouches, non-woven polypro / film, very resistant, suitable for STEAM 134°C, EO, FORM and PLASMA, cleanliness level 200 per IEST-STD-CC1246, 32x59cm, DIN EN ISO 11607, case of 5 packs x 100pcs 1 * 500 items
Catalog # VEPA3FKFB245140
Supplier: VP Medical Packaging
Frequently Bought Together