Microscope cover glasses, DIN ISO 8255 compliant, 22 mm Ø, /1000 borosilicate glass D 263Teco, thickness No. 3 (0.28-0.32 mm), CE ma rked, 50 pcs./two-part plastic box, 500 pcs./carton, 37.500 pcs./multipack, MARIENFELD SUPERIOR 1 * 1.000 items
Catalog # MARI0114620
Supplier: MARIENFELD
Frequently Bought Together