PHADEBAS AMYLASE TEST 1 * 50 ST
Catalog # MSPPPHADEBAS1301
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR® Amsterdam, Safety Spectacles
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
pH electrodes, SenTix® PLUS
Supplier: WTW
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Polyethylene oxide, M.W. >5,000,000
Supplier: Thermo Fisher Scientific
Bottles, Narrow Neck, with Screw Cap
Supplier: RIXIUS