Orders LinkContactUsLinkComponent
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (FITC)
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking slider
Catalog # MOTI1101011400191
Supplier:   MOTIC
FLED fluorescence intermediate attachments
Catalog # MOTI1101011400191
Supplier:  MOTIC
FLED fluorescence intermediate attachments
FLED fluorescence intermediate attachments
FLED Fluorescence intermediate attachment (FITC)
- Fluorescence intermediate part
- LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
- Light blocking slider
Price
1 950,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Compatible with Panthera U, KU and CC microscopes.
Spezifikationen
  • Description:
    FLED Fluorescence intermediate attachment (FITC)
    - Fluorescence intermediate part
    - LED excitation module FITC, consisting of 470 nm LED lamp and FITC filter cube
    - Light blocking slider