ContactUsLinkComponent
Complete splash head UDK129 the complete splash head includes the platic guard (10004708) and the standard test-tube connection (10002322) 1 * 1 items
Catalog # VELP40001624
Supplier:  VELP SCIENTIFIC
Complete splash head UDK129 the complete splash head includes the platic guard (10004708) and the standard test-tube connection (10002322) 1 * 1 items
Catalog # VELP40001624
Supplier:  VELP SCIENTIFIC
Frequently Bought Together