Order Entry
Austria
Orders LinkContactUsLinkComponent
Mop systems, HandySorb

Mop systems, HandySorb

Mop system 'No-Touch' universal pads, 47 L, 25 pads/case
Catalog # BRDY150598
Supplier:   Brady
Mop systems, HandySorb
Catalog # BRDY150598
Supplier:  Brady
Mop systems, HandySorb
Mop systems, HandySorb

Mop system 'No-Touch' universal pads, 47 L, 25 pads/case

Price
171,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

HandySorb is a unique and practical tool to safely remove spills in a highly efficient and easy way.

Safe spill clean-up: works with inert pads or pillows that can be replaced without ever touching the absorbed fluids
Easy to use: offers 1,83 m reach when fully extended and features a no touch snap-on system for pads
Highly efficient: each pad absorbs in seconds up to 25 times its weight

Specifications
  • Capacity:
    47 L
  • Description:
    Mop system 'No-Touch' universal pads
  • Seq:
    02
  • Packed:
    25 pads/case
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.