ContactUsLinkComponent
TLC Plastic Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent in dicator 1 * 25 items
  SIALZ740235-25EA
 :  Merck
TLC Plastic Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent in dicator 1 * 25 items
  SIALZ740235-25EA
 :  Merck
Frequently Bought Together