ContactUsLinkComponent
TLC Aluminum Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent i ndicator: no 1 * 25 items
Catalog # SIALZ740230-25EA
Supplier:  Merck
TLC Aluminum Sheets from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent i ndicator: no 1 * 25 items
Catalog # SIALZ740230-25EA
Supplier:  Merck
Frequently Bought Together