ContactUsLinkComponent
HPTLC (High Performance TLC) Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 10 cm x 10 cm, silica gel 60 matrix, binder, Po lymeric, fluorescent indicator: no 1 * 25 items
Catalog # SIALZ740222-25EA
Supplier:  Merck
HPTLC (High Performance TLC) Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 10 cm x 10 cm, silica gel 60 matrix, binder, Po lymeric, fluorescent indicator: no 1 * 25 items
Catalog # SIALZ740222-25EA
Supplier:  Merck
Frequently Bought Together