ContactUsLinkComponent
DRYSYN MULTI REACTION VIAL INSERTS, PACK OF 3 EACH INSERT IS 4 POSITION UNLESS OTHERWISE STATED. 11.80MM (WILL FIT 2ML HPLC SAMPLE V IALS) THIS SIZE IS A 9 POSITION INSERT. 1 * 3 items
Catalog # ASYNADS3-R-11.80
Supplier:  ASYNT
DRYSYN MULTI REACTION VIAL INSERTS, PACK OF 3 EACH INSERT IS 4 POSITION UNLESS OTHERWISE STATED. 11.80MM (WILL FIT 2ML HPLC SAMPLE V IALS) THIS SIZE IS A 9 POSITION INSERT. 1 * 3 items
Catalog # ASYNADS3-R-11.80
Supplier:  ASYNT
Frequently Bought Together