ContactUsLinkComponent
Mini-Desiccator Antistatic Premium, Black, static dissipative polycarbonate, including two shelves made of static dissipative polyet hylene, hygrometer and desiccant, non-slip rubber feet, stackable. All materials used can discharge electrostatic cha 1 * 1 items
Catalog # BOHLV1943-06
Supplier:  Bohlender
undefined
Mini-Desiccator Antistatic Premium, Black, static dissipative polycarbonate, including two shelves made of static dissipative polyet hylene, hygrometer and desiccant, non-slip rubber feet, stackable. All materials used can discharge electrostatic cha 1 * 1 items
Catalog # BOHLV1943-06
Supplier:  Bohlender
Frequently Bought Together