ContactUsLinkComponent
Star Desiccator Double, Aluminium frame with panels made of acrylic glass and double door, including four shelves made of stainless steel, tray, hygrometer and desiccant, usable with up to 26 shelves, stackable. 1 * 1 items
Catalog # BOHLV1930-07
Supplier:  Bohlender
undefined
Star Desiccator Double, Aluminium frame with panels made of acrylic glass and double door, including four shelves made of stainless steel, tray, hygrometer and desiccant, usable with up to 26 shelves, stackable. 1 * 1 items
Catalog # BOHLV1930-07
Supplier:  Bohlender
Frequently Bought Together