ContactUsLinkComponent
Multiple Distributors with Stopcock, Black screw cap made of PPS for bottle thread gl45, body made of PTFE. One stopcock for all con nectors. Very good chemical resistance. 1 * 1 items
Catalog # BOHLD617-08
Supplier:  Bohlender
undefined
Multiple Distributors with Stopcock, Black screw cap made of PPS for bottle thread gl45, body made of PTFE. One stopcock for all con nectors. Very good chemical resistance. 1 * 1 items
Catalog # BOHLD617-08
Supplier:  Bohlender
Frequently Bought Together