Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Plan fluorite oil; 1,3 NA, 0,2 mm
Catalog # ECHLJMAG0401
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # ECHLJMAG0401
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Plan fluorite oil; 1,3 NA, 0,2 mm
Price
5 630,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
    40×
  • Description:
    Plan fluorite oil; 1,3 NA
  • Seq:
    05
  • Working distance:
    0,2 mm