ContactUsLinkComponent
Flexible Tubing EX, Conductive corrugated tubing with nominal width 10 and with circular corrugations around the longitudinal axis. Cylindrical tubing ends with a length of 40 mm can for example be connected directly to fittings, stopcocks or hose co 1 * 1 items
Catalog # BOHLS1824-65
Supplier:  Bohlender
undefined
Flexible Tubing EX, Conductive corrugated tubing with nominal width 10 and with circular corrugations around the longitudinal axis. Cylindrical tubing ends with a length of 40 mm can for example be connected directly to fittings, stopcocks or hose co 1 * 1 items
Catalog # BOHLS1824-65
Supplier:  Bohlender
Frequently Bought Together