ContactUsLinkComponent
MDV 80031 Metalldetektor PA 6 - GF, 24 VDC, Relais Wechsler (Öffner), -25...+70°C, IP67,Kabel 2 m PVC [S80031] 1 * 1 ST
Catalog # EGEES80031
MDV 80031 Metalldetektor PA 6 - GF, 24 VDC, Relais Wechsler (Öffner), -25...+70°C, IP67,Kabel 2 m PVC [S80031] 1 * 1 ST
Catalog # EGEES80031
Frequently Bought Together