Submersible pump set (low cost). Complete set for monitoring wells O 40 mm, consisting of submersible pump Gigant (3x), booster pump (3x), extension cable, sample tube and battery with battery-charger. For sampling till a depth of 14.5 m. 1 * 1 SET
Catalog # EIJK1212SA
Supplier: EIJKELKAMP AGRISECTOR
Frequently Bought Together