O-ring 3475 viton 1 * 1 items
Catalog # VELP10002033
Supplier: VELP SCIENTIFIC
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for accu-jet® pro
Supplier: Brand
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for water purification millipore
Supplier: Merck