Order Entry
Austria
Orders LinkContactUsLinkComponent
Dextran 40 USP

Dextran 40 USP

1 kg
Catalog # SPCMD2450-1KGBL
Supplier:   Spectrum Chemical
Dextran 40 USP
Catalog # SPCMD2450-1KGBL
Supplier:  Spectrum Chemical
CAS Number:  9004-54-0
Dextran 40 USP

1 kg

CAS Number:  9004-54-0
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents

Dextran 40, Powder, USP is used as an antithrombotic in order to reduce blood viscosity. It is a complex branched glucan that is synthethised by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.