SWAB SOFTA CLEANSING SWAB ININTIN 1 * 100 items
Catalog # BRAU19579
Supplier: B. Braun
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Surface disinfectant, Meliseptol® rapid
Supplier: B. Braun
Calcium carbonate Certipur Reag. USP 1 * 50 g
Supplier: Merck
Accessories for Sartorius printers
Supplier: Sartorius Balances
Cell scrapers and cell lifters, Corning®
Supplier: Corning
Insulin syringes, BD Micro-Fine™
Supplier: EMBECTA
Sharpsafe® Ecological 5th Generation Recycled Needle Containers
Supplier: Hospidex
Quantitative filter papers, ashless grades, grade 589/3, Whatman™
Supplier: Whatman products (Cytiva)
Sulphuric acid ≥96%, Supelco®
Supplier: Merck
Eyewash solution
Supplier: Plum Safety
Safety needles, Sterican®
Supplier: B. Braun
ESD safety shoes, lace-up, uvex1, 8544
Supplier: UVEX
Buffer solution pH 9.180 @25?C NIST and DIN 19266,6 x 250ml, Mettler 1 * 6 items
Supplier: Mettler - Toledo
ESD safety ankle boots, lace-up, L10, Emotion
Supplier: ELTEN
Acetic acid 25% in water
Supplier: Thermo Fisher Scientific
Accessories for GRINDOMIX GM 200
Supplier: Retsch GmbH
(±)-1,2-Propanediol, liquid
Supplier: MP Biomedicals
Safety shoes, slip-on, Uni6, 1740
Supplier: Abeba