CLOSED TOP SCREW CAPS FOR 4ML 15X45MM13-425MM THREADED AUTOSAMPLER VIALS WITHOUT SEPTA (SOLD SEPARATELY). MICROSOLV BRAND. 1 * 100 items
Catalog # MICS9504S-01C-BK
Supplier: MICROSOLV
CLOSED TOP SCREW CAPS FOR 4ML 15X45MM13-425MM THREADED AUTOSAMPLER VIALS WITHOUT SEPTA (SOLD SEPARATELY). MICROSOLV BRAND. 1 * 100 items
Catalog # MICS9504S-01C-BK
Supplier: MICROSOLV
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Magnetic stirring bars, micro
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals