Order Entry
Austria
Orders LinkContactUsLinkComponent
epT.I.P.S.® Motion SafeRack Tips

epT.I.P.S.® Motion SafeRack Tips

PCR clean, 50 µl, -, +, 10 racks of 96 tips
Catalog # EPPE0030014.618
Supplier:   EPPENDORF
epT.I.P.S.® Motion SafeRack Tips
Catalog # EPPE0030014.618
Supplier:  EPPENDORF
epT.I.P.S.® Motion SafeRack Tips
epT.I.P.S.® Motion SafeRack Tips

PCR clean, 50 µl, -, +, 10 racks of 96 tips

Price
244,00 €
Log in for availability
1 * 960 ST
Product Details & Documents

The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well. Here special epT.I.P.S. Motion SafeRack tips are recommended.

  • Colour-coded trays for direct volume identification
  • Double O-ring on dispensing tools assures an optimal fit each time
  • Optical sensor automatically identified tip type
  • Box design with internal, individual tip compartments to avoid cross-contamination between used tips
  • Available as standard and PCR clean quality for 50, 300 and 1000 µl

Pipette tips will be placed back into the position of the box they were removed from. Returned tips will be reused within an application for the same source vessel.

Automated pipetting processes place high demands on the design and material of pipette tips used. Eppendorf has specially designed the epT.I.P.S. for use on the epMotion® 5070 and 5075 Liquid handling workstation. epT.I.P.S. Motion SafeRack pipette tips, with and without filters, developed in torsion-resistant, contamination-free racks that are recommended for the re-use of tips feature of epMotion 5073 systems.

Applications
- Available in multiple purity grades.
- Easily exchangeable, no additional labware files required.

Specifications
  • Packed:
    10 racks of 96 tips
  • Sterile:
    -
  • Volume:
    50 µl
  • Seq:
    02
  • Tip style:
    PCR clean
  • Filter:
    +
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.