PROTEINASE K SOLUTION 25ML (20MG/ML) GENOMIC PCR GRADE PROTEINASE K SOLUTIO
Catalog # VIAG503-PK
Supplier: VIAGEN BIOTECH
PROTEINASE K SOLUTION 25ML (20MG/ML) GENOMIC PCR GRADE PROTEINASE K SOLUTIO
Catalog # VIAG503-PK
Supplier: VIAGEN BIOTECH
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microscope Slides
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Label Dots on Rolls
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
DirectPCR® DNA extraction system
Supplier: VIAGEN BIOTECH
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
VWR® Reagent Reservoirs
Supplier: VWR Collection