Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tube centrifuge, 50 ml, blue graduated, scale up to 20 ml and mark at 50 ml,for determination of solubility acc. to ADPI 1 * 10 items
Catalog # MARI3949501
Supplier:
MARIENFELD
Expand image
Tube centrifuge, 50 ml, blue graduated, scale up to 20 ml and mark at 50 ml,for determination of solubility acc. to ADPI 1 * 10 items
Catalog # MARI3949501
Supplier:
MARIENFELD
Frequently Bought Together
VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
Macro Cells, Quartz Glass for Absorption Measurements
Supplier: HELLMA
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Screw caps for laboratory bottles, DURAN®
Supplier: DWK Life Sciences
Beakers, low form, DURAN®
Supplier: DWK Life Sciences
Round cuvettes
Supplier: Hach