COUNT-CAL COUNT PRECISION SIZE STANDARDS-6 X 25ML 1 * 150 mL
Catalog # MSPPCC15-PK
Supplier: MARKET SOURCE PART PROCESS
COUNT-CAL COUNT PRECISION SIZE STANDARDS-6 X 25ML 1 * 150 mL
Catalog # MSPPCC15-PK
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Formic acid ≥99%
Supplier: Thermo Fisher Scientific
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
DISTRILAB Count-Cal Count Precision Standard (6x25ml) 1 * 150 mL
Supplier: MARKET SOURCE PART PROCESS
Accessories for CO₂ incubators, MCO-19AIC series, PHCbi
Supplier: PHC Corporation