Volumetric flask, class a ray-sorb® heavy duty wide mouth with ptfe stopper, capacity (ml)-50, tolerance (ml)±0.08, height includin g stopper (mm)-147, standard taper stopper-13, stopper color-orange. 1 * 1 items
Catalog # SCERSP17810-19300
Supplier: DWK Life Sciences
Frequently Bought Together