ContactUsLinkComponent
5ml disposable tip for LiHa Arm. SBS footprint rack compatible with standard carriers. 10 racks / 240 tips. Tecan Pure: certified free of human DNA, RNase, DNase and PCR inhibitors
Catalog # TCAN30059897
Supplier:  Tecan
5ml disposable tip for LiHa Arm. SBS footprint rack compatible with standard carriers. 10 racks / 240 tips. Tecan Pure: certified free of human DNA, RNase, DNase and PCR inhibitors
Catalog # TCAN30059897
Supplier:  Tecan
Frequently Bought Together