ContactUsLinkComponent
Dispenser ceramus 0.2-1.0ml conformity certifiedPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
Catalog # HIRS9312001
Supplier:  HIRSCHMANN
Dispenser ceramus 0.2-1.0ml conformity certifiedPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
Catalog # HIRS9312001
Supplier:  HIRSCHMANN
Frequently Bought Together