ContactUsLinkComponent
: Peelable heat sealing foil, suitable for very low temperature storage/high temperature uses/PCR, roll (610 m x 78 mm, approx. 5,00 0 seals
Catalog # FTIT4TI-0520
Supplier:  4titude Ltd.
: Peelable heat sealing foil, suitable for very low temperature storage/high temperature uses/PCR, roll (610 m x 78 mm, approx. 5,00 0 seals
Catalog # FTIT4TI-0520
Supplier:  4titude Ltd.
Frequently Bought Together