J&W CP-Wax 52 CB, 60 m, 0.32 mm, 0.25 µm, GC column, 7 inch cage 1 * 1 items
Catalog # HEWLCP8853I
Supplier: Agilent
J&W CP-Wax 52 CB, 60 m, 0.32 mm, 0.25 µm, GC column, 7 inch cage 1 * 1 items
Catalog # HEWLCP8853I
Supplier: Agilent
Frequently Bought Together
Air valves, b.safe
Supplier: Bohlender
VWR®, Snap Ring Vials ND11
Supplier: VWR Collection
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
Vydac 218TP C18, HPLC columns
Supplier: Avantor
VWR®, Storage Boxes for Sample Vials and Bottles, PP
Supplier: VWR Collection
b.safe Cap GL 45 UHPLC
Supplier: Bohlender
Caps for solvent supplies, b.safe
Supplier: Bohlender
Capillary columns, OPTIMA WAXplus®
Supplier: MACHEREY-NAGEL
Tubing, flanged, PTFE, BOLA
Supplier: Bohlender
Filters for advantage 200 LS
Supplier: M S A
SAMPLE RACK 9+29 POS. FOR AS93 PLUS 1 * 1 items
Supplier: PerkinElmer
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI
Waste caps, b.safe
Supplier: Bohlender
Inlet septa
Supplier: Agilent
Exhaust filters, b.safe
Supplier: Bohlender
Accessories for HV valves
Supplier: HAMILTON BONADUZ
Capillaries for LC systems
Supplier: Agilent
Avantor® Prevail Organic acid, HPLC Columns
Supplier: Avantor