ContactUsLinkComponent
Maintenance ISO-11171 Calibration for 20-1111 PAMAS SBSS Particle counter - check of optical components / alignment - check of electronics / alignment - check of hydraulic flowpath (alignment work, if necessary, will be charged separately) includes b 1 * 1 ST
Catalog # IBSAISO-11171
Supplier:  IBS Stock Items
Maintenance ISO-11171 Calibration for 20-1111 PAMAS SBSS Particle counter - check of optical components / alignment - check of electronics / alignment - check of hydraulic flowpath (alignment work, if necessary, will be charged separately) includes b 1 * 1 ST
Catalog # IBSAISO-11171
Supplier:  IBS Stock Items
Frequently Bought Together