Orders LinkContactUsLinkComponent
Reading magnifiers, visoPOCKET
Reading magnifiers, visoPOCKET
Reading magnifier, brown, visoPOCKET, 74×48 mm
Catalog # ESCH172146
Supplier:   ESCHENBACH OPTIK
Reading magnifiers, visoPOCKET
Catalog # ESCH172146
Supplier:  ESCHENBACH OPTIK
Reading magnifiers, visoPOCKET
Reading magnifiers, visoPOCKET
Reading magnifier, brown, visoPOCKET, 74×48 mm
Price
36,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Compact magnifiers with wide field of view and ultra-slim lenses.
  • Diffractive aspheric lens with modern and exquisite design
  • High quality leather available in three colours
  • Leather case protects from scratches
Sutiable for on-the-go use when small print needs magnifying swiftly and easily.
Specifications
  • Magnification:
    2,5×
  • Description:
    Reading magnifier, brown, visoPOCKET
  • W×D:
    74×48 mm