Reading magnifiers, visoPOCKET
Reading magnifier, red, visoPOCKET, 74×48 mm
Catalog # ESCH172116
Supplier: ESCHENBACH OPTIK
Springe zu:
Product Details & Documents
Compact magnifiers with wide field of view and ultra-slim lenses.
- Diffractive aspheric lens with modern and exquisite design
- High quality leather available in three colours
- Leather case protects from scratches
Sutiable for on-the-go use when small print needs magnifying swiftly and easily.
Spezifikationen
- Magnification:2,5×
- Description:Reading magnifier, red, visoPOCKET
- W×D:74×48 mm