Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
2-way header with ball valves, M30×1,5, The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Catalog # HUBE343314
Supplier:   HUBER
Accessories for circulators
Catalog # HUBE343314
Supplier:  HUBER
Accessories for circulators
2-way header with ball valves, M30×1,5, The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Price
760,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    2-way header with ball valves, M30×1,5
  • Seq:
    08
  • For:
    The 2-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.