Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
2-way header, M16×1, The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
Catalog # HUBE337657
Supplier:   HUBER
Accessories for circulators
Catalog # HUBE337657
Supplier:  HUBER
Accessories for circulators
2-way header, M16×1, The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.
Price
275,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    2-way header, M16×1
  • Seq:
    01
  • For:
    The 2-way header and the TCS should be connected with a suitable length of insulated hose. Universal mounting device for fixing in position is included. For direct connection to the unit, an additional elbow connector is required.