ContactUsLinkComponent
Chilltech regional 10 litre system 2-8C, complete Kit. 1 * 1 items
Catalog # SCATCHR10U
Supplier:  Sonoco Thermosafe
Chilltech regional 10 litre system 2-8C, complete Kit. 1 * 1 items
Catalog # SCATCHR10U
Supplier:  Sonoco Thermosafe
Frequently Bought Together