ContactUsLinkComponent
G-Sep Agarose 6B is a gel filtration matrix formed from agarose beads that is available with 6% agarose content, designated G-Sep Agarose 6B.The resin is a proven gel filtration base matrix and is routinely modified by researchers to couple affinit 1 * 100 mL
Catalog # GENO786-1568
Supplier:  G-Biosciences
G-Sep Agarose 6B is a gel filtration matrix formed from agarose beads that is available with 6% agarose content, designated G-Sep Agarose 6B.The resin is a proven gel filtration base matrix and is routinely modified by researchers to couple affinit 1 * 100 mL
Catalog # GENO786-1568
Supplier:  G-Biosciences
Frequently Bought Together