Dimethylsulfoxide (DMSO) is a very polar organic compound which used for solubilizing organic and inorganic compounds. In addition, DMSO is used in cell culture as a cryoprotectant added to cell culture medium, to prevent formation of ice crystals d 1 * 50 mL
Catalog # GENO786-1388
Supplier: G-Biosciences
Frequently Bought Together