Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Thermal transfer printer labels, type B-499

Printer labels, B-499, BPT-603-499YL, Nylon, Permanent acrylic, Thermal transfer, Yellow, BBP12, 12 mm, 3000 labels/roll
Catalog # BRDY217069
Supplier:   Brady
Thermal transfer printer labels, type B-499
Catalog # BRDY217069
Supplier:  Brady
Thermal transfer printer labels, type B-499
Thermal transfer printer labels, type B-499

Printer labels, B-499, BPT-603-499YL, Nylon, Permanent acrylic, Thermal transfer, Yellow, BBP12, 12 mm, 3000 labels/roll

Price
265,00 €
Log in for availability
1 * 1 Rolle
Product Details & Documents

Printable, nylon cloth labels for thermal transfer printers.

  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves

This material is UL recognised with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.

Specifications
  • Printer model:
    BBP12
  • Printing method:
    Thermal transfer
  • Type:
    B-499
  • Colour:
    Yellow
  • Description:
    Printer labels
  • Dimensions:
    12 mm
  • Material:
    Nylon
  • Seq:
    07
  • Model:
    BPT-603-499YL
  • Packed:
    3000 labels/roll
  • Adhesive:
    Permanent acrylic