E. coli OP50 (for C. elegans) 5 beads. Store desiccated in the refrigerator. 1 * 5 items
Catalog # EDVO729
Supplier: EDVOTEK
E. coli OP50 (for C. elegans) 5 beads. Store desiccated in the refrigerator. 1 * 5 items
Catalog # EDVO729
Supplier: EDVOTEK
Frequently Bought Together
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Dichloromethane stabilised Ph. Eur.
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexanol for synthesis
Supplier: Merck
Diisodecyl phthalate, Sigma-Aldrich®
Supplier: Merck
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
NEMATODE GROWTH MEDIUM (NGM) POWDER 1 * 1 kg
Supplier: US Biological
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals