ContactUsLinkComponent
(1) 4-20% Tris-Glycine-SDS Precast Polyacrylamide Gel (PAGE) , Size: 9 x 10 cm , Designed for Separating Protein Fragments 20-100 kD a , Each Gel Has 10 Wells , Well Volume is 30 µl, Requires Refrigeration. Use within 3 months of receipt 1 * 1 items
Catalog # EDVO650
Supplier:  EDVOTEK
(1) 4-20% Tris-Glycine-SDS Precast Polyacrylamide Gel (PAGE) , Size: 9 x 10 cm , Designed for Separating Protein Fragments 20-100 kD a , Each Gel Has 10 Wells , Well Volume is 30 µl, Requires Refrigeration. Use within 3 months of receipt 1 * 1 items
Catalog # EDVO650
Supplier:  EDVOTEK
Frequently Bought Together