ContactUsLinkComponent
This powdered blend Tris-Borate EDTA buffer (TBE), has been optimized for agarose gel electrophoresis of nucleic acids. Dissolve pow der in 500 ml of distilled or deionized water to prepare a 10x concentrate which can be diluted 10-fold to make a 1x 1 * 1 items
Catalog # EDVO607-1
Supplier:  EDVOTEK
This powdered blend Tris-Borate EDTA buffer (TBE), has been optimized for agarose gel electrophoresis of nucleic acids. Dissolve pow der in 500 ml of distilled or deionized water to prepare a 10x concentrate which can be diluted 10-fold to make a 1x 1 * 1 items
Catalog # EDVO607-1
Supplier:  EDVOTEK
Frequently Bought Together