ContactUsLinkComponent
Dissolved oxygen levels are used to monitor the health and productivity of aquatic ecosystems. In this kit, students familiarize the mselves with the Winkler method by examining the relationship between temperature and dissolved oxygen concentrations 1 * 1 KIT
Catalog # EDVO292
Supplier:  EDVOTEK
Dissolved oxygen levels are used to monitor the health and productivity of aquatic ecosystems. In this kit, students familiarize the mselves with the Winkler method by examining the relationship between temperature and dissolved oxygen concentrations 1 * 1 KIT
Catalog # EDVO292
Supplier:  EDVOTEK
Frequently Bought Together