ContactUsLinkComponent
MiniOne PCR system with Android mobile tablet. Includes thermal cycler with 16 sample well capacity. Fast heating & cooling based on peltier-effect thermoelectric, heated lid, wireless connectivity to iPad or Android mobile tablets. Includes 7 mobi 1 * 1 items
Catalog # EMBIM4001
Supplier:  EMBITEC
MiniOne PCR system with Android mobile tablet. Includes thermal cycler with 16 sample well capacity. Fast heating & cooling based on peltier-effect thermoelectric, heated lid, wireless connectivity to iPad or Android mobile tablets. Includes 7 mobi 1 * 1 items
Catalog # EMBIM4001
Supplier:  EMBITEC
Frequently Bought Together