Acidic cleaner deconex 41 LAB CLEAR 1 * 25 kg
Catalog # DECO503715.00-K25R
Lieferant: Borer Chemie
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Lieferant: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Lieferant: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Lieferant: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Lieferant: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Lieferant: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Lieferant: VWR Chemicals
Disinfectant solutions, Bacillol AF
Lieferant: BODE CHEMIE