Orders LinkContactUsLinkComponent
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
Grey
  479-2486
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
  479-2486
   Low Manufacturing Impact
Colour coders for cryovials, CryoColorCode, Nunc™
Colour coders for cryovials, CryoColorCode, Nunc™
Grey
18,00 €
  30/06/2026
1 * 500 ST
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
  • Colour:
    Grey
  • Seq:
    13
  • Cat. No.:
    479-2486
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact