ContactUsLinkComponent
Stericlin® sterilisation pouches, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, 32 x 45 cm, DIN EN ISO 11607 1 * 500 items
Catalog # 129-1290
Stericlin® sterilisation pouches, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, 32 x 45 cm, DIN EN ISO 11607 1 * 500 items
Catalog # 129-1290