ContactUsLinkComponent
Stericlin Bowie-Dick-Helix test, contains 1 aluminium Process Challenge Device (can be used for 250 cycles) and 250 single-use test strips, DIN EN ISO 11140-
Catalog # 489-0118
Stericlin Bowie-Dick-Helix test, contains 1 aluminium Process Challenge Device (can be used for 250 cycles) and 250 single-use test strips, DIN EN ISO 11140-
Catalog # 489-0118